Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3830406C13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284383.1 | NP_001271312.1 | 5 | 442 | ||
NM_001284384.1 | NP_001271313.1 | 5 | 518 | |||
NM_001284385.1 | NP_001271314.1 | 5 | 521 | |||
NM_001284386.1 | NP_001271315.1 | 4 | 361 | |||
NM_146051.3 | NP_666163.1 | 6 | 506 | |||
NM_178141.3 | NP_835277.1 | 5 | 445 | |||
GenBank mRNA | AI931548.1 | - | 3 | 239 | ||
AK076182.1 | - | 5 | 436 | |||
AK078647.1 | - | 1 | 886 | |||
AK089971.1 | - | 5 | 418 | |||
BC027421.1 | - | 5 | 466 | 4194414 | ||
BC057656.1 | - | 5 | 467 | 4020532 | ||
BC094456.1 | - | 5 | 423 | 5137611 | ||
BQ715343.1 | - | 5 | 464 |