Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181404.5 | NP_852069.4 | 8, 9 | 3677 | ||
GenBank mRNA | AK084638.1 | - | 1, 2 | 127 | ||
AK122214.1 | - | 4, 5 | 2043 | |||
AK147945.1 | - | 5, 6 | 3264 | |||
BC050250.1 | - | 3, 4 | 484 | |||
BC079563.1 | - | 4, 5 | 3004 | 30535512 |