Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nppc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010933.5 | NP_035063.1 | 2 | 279 | ||
GenBank mRNA | AK020093.1 | - | 2 | 282 | ||
AK036176.1 | - | 2 | 281 | |||
BC111834.1 | - | 2 | 210 | 40055241 | ||
BY718381.1 | - | 2 | 280 |