Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sprr2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164787.1 | NP_001158259.1 | 284 | |||
GenBank mRNA | AK007928.1 | - | 195 | |||
BC010818.1 | - | 184 | 4216389 |
Gene Symbol: Sprr2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011468.4 | NP_035598.2 | 2 | 284 | ||
GenBank mRNA | AK003391.1 | - | 2 | 200 | ||
AK007633.1 | - | 2 | 200 | |||
AK008631.1 | - | 2 | 197 | |||
AK008887.1 | - | 2 | 200 | |||
AK009238.1 | - | 2 | 199 | |||
AK009790.1 | - | 2 | 197 | |||
AK147183.1 | - | 2 | 200 | |||
AK168529.1 | - | 2 | 199 | |||
AY158986.1 | - | 197 | ||||
BY062071.1 | - | 2 | 309 |
Gene Symbol: Sprr2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC051122.1 | - | 1 | 159 | ||
BC056231.1 | - | 1 | 170 | |||
BC099393.1 | - | 1 | 188 | 30790331 | ||
BY094335.1 | - | 196 |