Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278581.1 | NP_001265510.1 | 4 | 822 | ||
NM_001278582.1 | NP_001265511.1 | 4 | 757 | |||
NM_001278583.1 | NP_001265512.1 | 4 | 727 | |||
NM_001278584.1 | NP_001265513.1 | 3 | 616 | |||
NM_008895.4 | NP_032921.1 | 3 | 646 | |||
GenBank mRNA | AK017492.1 | - | 3 | 642 | ||
AK017581.1 | - | 3 | 611 | |||
AK030714.1 | - | 4 | 721 | |||
AK077426.1 | - | 3 | 610 | |||
AK133592.1 | - | 3 | 641 | |||
AK133646.1 | - | 3 | 641 | |||
AK133740.1 | - | 3 | 640 | |||
AK133776.1 | - | 3 | 642 | |||
AK133800.1 | - | 3 | 642 | |||
BB022229.2 | - | 1 | 302 | |||
BC061215.1 | - | 3 | 640 | 30253829 |