Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033596.3 | NP_291074.1 | 1 | 261 | ||
GenBank mRNA | BC051566.1 | - | 1 | 269 | ||
BC057955.1 | - | 255 | 5359141 | |||
BX520915.1 | - | 1 | 261 |