Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lce1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026822.1 | NP_081098.1 | 2 | 530 | ||
GenBank mRNA | AK003951.1 | - | 2 | 530 | ||
BC069979.1 | - | 2 | 507 | |||
BC082291.1 | - | 2 | 502 | |||
BC132596.1 | - | 1 | 481 | 40130873 | ||
BC132598.1 | - | 1 | 481 | 40130875 |