Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dolk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177648.3 | NP_808316.1 | 1 | 1044 | ||
GenBank mRNA | AB073968.1 | - | 1 | 987 | ||
AK077581.1 | - | 1 | 1014 | |||
AK144705.1 | - | 1 | 1025 | |||
BC019543.1 | - | 1 | 989 | |||
BC026973.1 | - | 1 | 769 | 5371267 |