Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2a1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007535.2 | NP_031561.2 | 1 | 179 | ||
GenBank mRNA | BC120720.1 | - | 1 | 179 | 40129643 | |
BC132545.1 | - | 1 | 179 |