Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ins2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185083.1 | NP_001172012.1 | 3 | 327 | ||
NM_001185084.1 | NP_001172013.1 | 2 | 441 | |||
NM_008387.4 | NP_032413.1 | 3 | 339 | |||
GenBank mRNA | AK007612.1 | - | 3 | 340 | ||
AY899305.1 | - | 2 | 406 | |||
BC066208.1 | - | 3 | 348 | |||
BC099934.1 | - | 3 | 312 | 6432976 | ||
BC132650.1 | - | 3 | 312 | 40130927 | ||
BC132652.1 | - | 3 | 312 | 40130929 | ||
BC145554.1 | - | 3 | 300 | 9054118 | ||
GQ915612.1 | - | 3 | 327 |