Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021358.2 | NP_067333.1 | 2 | 816 | ||
GenBank mRNA | AF134158.1 | - | 2 | 816 | ||
BC116652.1 | - | 2 | 816 | 40104888 | ||
BC116653.1 | - | 2 | 816 | 40104889 |
Set Membership: |
![]() |