- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Gene Symbol: 1110006O24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB041800.1 | - | 1 | 452 | ||
AK003508.1 | - | 1 | 453 | |||
AK006095.1 | - | 1 | 423 | |||
BC049589.1 | - | 1 | 432 | |||
BC061108.1 | - | 1 | 443 |