Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013550.5 | NP_038578.2 | 1 | 443 | ||
GenBank mRNA | AK006742.1 | - | 1 | 423 | ||
AK018952.1 | - | 1 | 444 | |||
BC115816.1 | - | 1 | 440 | 40105082 |