Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023680.3 | NP_076169.2 | 3 | 289 | ||
GenBank mRNA | AJ278265.1 | - | 3 | 247 | ||
AK012838.1 | - | 3 | 290 | |||
AY046551.1 | - | 3 | 288 | |||
AY165626.1 | - | 3 | 312 | |||
AY165627.1 | - | 3 | 312 | |||
BC076592.1 | - | 3 | 303 |