Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013560.2 | NP_038588.2 | 1, 2 | 517 | ||
GenBank mRNA | AF047377.1 | - | 2, 3 | 210 | ||
AF047378.1 | - | 2, 3 | 134 | |||
AK003119.1 | - | 1, 2 | 438 | |||
AK010663.1 | - | 1, 2 | 435 | |||
AK168927.1 | - | 1, 2 | 435 | |||
BC018257.1 | - | 1, 2 | 457 | 3498180 | ||
BC090388.1 | - | 1, 2 | 418 | |||
BC099463.1 | - | 1, 2 | 412 | 30848584 | ||
U03560.1 | - | 1, 2 | 374 | |||
U03561.1 | - | 2, 3 | 341 | |||
U03562.1 | - | 2, 3 | 275 | |||
X14686.1 | - | 1, 2 | 419 | |||
X14687.1 | - | 1, 2 | 464 |