Gene Symbol: Pde6h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023898.4 | NP_076387.2 | 3, 4 | 275 | ||
GenBank mRNA | AF190929.1 | - | 2, 3 | 183 | ||
AK020922.1 | - | 3, 4 | 276 | |||
AK080696.1 | - | 3, 4 | 291 | |||
BC057290.1 | - | 247 | ||||
BC058192.1 | - | 2, 3 | 2035 | |||
BC115671.1 | - | 3, 4 | 253 | 40104107 | ||
BC115672.1 | - | 3, 4 | 253 | 40104108 |