Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap3-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025524.2 | NP_079800.1 | 1 | 462 | ||
GenBank mRNA | AK009052.1 | - | 1 | 466 | ||
AK009410.1 | - | 1 | 464 | |||
AK010142.1 | - | 1 | 463 | |||
BC120760.1 | - | 1 | 430 | 40129683 | ||
BC120762.1 | - | 1 | 430 | 40129685 |