Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026278.3 | NP_080554.1 | 2 | 309 | ||
GenBank mRNA | AK007202.1 | - | 2 | 301 | ||
AK015594.1 | - | 2 | 302 | |||
AK016626.1 | - | 314 | ||||
BC051150.1 | - | 2 | 309 | 6772814 |