Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam83h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168253.1 | NP_001161725.1 | 2 | 269 | ||
NM_134087.2 | NP_598848.2 | 2 | 237 | |||
GenBank mRNA | AK030392.1 | - | 2 | 270 | ||
AK035531.1 | - | 2 | 237 | |||
BC117947.1 | - | 2 | 210 | 40098049 | ||
BC117948.1 | - | 2 | 210 | 40098059 |