Gene Symbol: STK32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018401.2 | NP_060871.1 | 2, 3 | 449 | ||
XM_011513497.1 | XP_011511799.1 | 2, 3 | 643 | |||
XM_017008374.1 | XP_016863863.1 | 1, 2 | 50 | |||
XM_017008375.1 | XP_016863864.1 | 1, 2 | 50 | |||
XM_017008376.1 | XP_016863865.1 | 1, 2 | 50 | |||
GenBank mRNA | AJ250839.1 | - | 2, 3 | 166 | ||
AK292696.1 | - | 2, 3 | 416 | |||
AK313846.1 | - | 2, 3 | 451 | |||
AY358353.1 | - | 2, 3 | 193 | |||
BC038238.1 | - | 2, 3 | 143 | 5744969 |