Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001677.3 | NP_001668.1 | 2, 3 | 355 | ||
GenBank mRNA | AK290181.1 | - | 2, 3 | 368 | ||
AK296059.1 | - | 2, 3 | 243 | |||
AK316043.1 | - | 2, 3 | 300 | |||
AY545986.1 | - | 2, 3 | 234 | |||
BC000006.2 | - | 2, 3 | 320 | 3506311 | ||
BQ934151.1 | - | 3, 4 | 559 | |||
BT009787.1 | - | 2, 3 | 234 | |||
CR407616.1 | - | 2, 3 | 234 | |||
U16799.1 | - | 2, 3 | 279 |