Gene Symbol: Olfr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146964.1 | NP_667175.1 | 1 | 618 | ||
GenBank mRNA | AF102540.1 | - | 1 | 426 | ||
BC119308.1 | - | 1 | 714 | 8734057 | ||
BC119334.1 | - | 1 | 714 | 8734083 |