Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001700.4 | NP_001691.1 | 4 | 532 | ||
GenBank mRNA | BC069495.1 | - | 4 | 527 | 7262247 | |
BC093931.1 | - | 4 | 532 | 7939776 |