Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033929.2 | NP_001029101.1 | 9 | 1622 | ||
NM_178413.5 | NP_848500.3 | 10 | 1690 | |||
GenBank mRNA | AK165463.1 | - | 9 | 1588 | ||
BC025604.1 | - | 10 | 1653 | 5250319 | ||
BC051244.1 | - | 9 | 1584 | 4975661 |