Search Thermo Fisher Scientific
Gene Symbol: Fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166368.1 | NP_001159840.1 | 2 | 170 | ||
NM_008020.3 | NP_032046.1 | 2 | 185 | |||
GenBank mRNA | AK144284.1 | - | 2 | 186 | ||
AK165394.1 | - | 2 | 150 | |||
BC031824.1 | - | 2 | 139 | 4196363 | ||
BC053692.1 | - | 2 | 175 | 6395841 | ||
DV655383.1 | - | 2 | 170 |