Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010858.4 | NP_034988.2 | 3, 4 | 272 | ||
GenBank mRNA | AK013098.1 | - | 1, 2 | 200 | ||
BC048485.1 | - | 3, 4 | 272 | 6741975 |