Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retnla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020509.3 | NP_065255.2 | 2 | 185 | ||
GenBank mRNA | AF205951.1 | - | 1 | 118 | ||
AF290872.1 | - | 2 | 223 | |||
AF316397.2 | - | 2 | 127 | |||
AF323082.1 | - | 2 | 195 | |||
AK007553.1 | - | 2 | 130 | |||
BC029248.1 | - | 1 | 125 | 4189285 |