Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111025.1 | NP_001104495.1 | 4 | 619 | ||
NM_024464.3 | NP_077784.2 | 4 | 625 | |||
GenBank mRNA | AK008583.2 | - | 4 | 627 | ||
BC002202.1 | - | 2 | 269 | 3489499 | ||
BY708475.1 | - | 4 | 626 |