Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025957.4 | NP_080233.1 | 2 | 195 | ||
GenBank mRNA | AK005390.1 | - | 2 | 196 | ||
AK005482.1 | - | 2 | 196 | |||
AK005533.1 | - | 2 | 196 | |||
BC099412.1 | - | 2 | 125 | 30787387 |