Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026061.2 | NP_080337.1 | 1, 2 | 105 | ||
GenBank mRNA | AK010177.1 | - | 1, 2 | 95 | ||
AK013516.1 | - | 1, 2 | 105 | |||
AK028070.1 | - | 1, 2 | 116 | |||
BC043019.1 | - | 1, 2 | 105 | 5684256 |