Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rpl22l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026517.2 | NP_080793.1 | 2 | 102 | ||
GenBank mRNA | AK008915.1 | - | 2 | 104 | ||
AK010265.1 | - | 2 | 83 | |||
AK013966.1 | - | 2 | 102 | |||
AK170359.1 | - | 1 | 1062 | |||
BC026533.1 | - | 2 | 82 | 5030430 | ||
BC120716.1 | - | 2 | 177 | 40129639 | ||
BC120718.1 | - | 2 | 177 | 40129641 |
Gene Symbol: Gm15421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK331818.1 | - | 1 | 420 |