Gene Symbol: Smim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133697.3 | NP_598458.1 | 4 | 282 | ||
GenBank mRNA | AK003345.1 | - | 4 | 277 | ||
AK018994.1 | - | 4 | 291 | |||
AK044669.1 | - | 4 | 386 | |||
AK088833.1 | - | 4 | 329 | |||
AK164394.1 | - | 4 | 283 | |||
BC009097.1 | - | 4 | 277 | 3481841 |