Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133779.2 | NP_598540.2 | 1 | 148 | ||
GenBank mRNA | AK019717.2 | - | 1 | 139 | ||
AK044499.1 | - | 1 | 132 | |||
AK049171.1 | - | 1 | 141 | |||
AK049990.1 | - | 1 | 141 | |||
AK090020.1 | - | 1 | 141 | |||
AK153462.1 | - | 1 | 134 | |||
AK153464.1 | - | 1 | 134 | |||
AK155057.1 | - | 1 | 135 | |||
AK157226.1 | - | 1 | 131 | |||
AK162573.1 | - | 1 | 128 | |||
BC016520.1 | - | 1 | 112 | |||
BC034175.1 | - | 1 | 124 | 5026752 | ||
BU058874.1 | - | 1 | 148 |