Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291049.1 | NP_001277978.1 | 1, 2 | 609 | ||
NM_173754.4 | NP_776115.2 | 1, 2 | 609 | |||
GenBank mRNA | AK042217.1 | - | 1, 2 | 609 | ||
AK083271.1 | - | 1, 2 | 447 |