Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003852.3 | NP_003843.3 | 2, 3 | 706 | ||
NM_015905.2 | NP_056989.2 | 2, 3 | 706 | |||
GenBank mRNA | AF009353.1 | - | 2, 3 | 479 | ||
AF119042.1 | - | 2, 3 | 607 | |||
AK075306.1 | - | 2, 3 | 706 | |||
AK302679.1 | - | 3, 4 | 579 | |||
AK312610.1 | - | 2, 3 | 834 | |||
BC028689.2 | - | 2, 3 | 604 | 4827559 |