Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001662.3 | NP_001653.1 | 6 | 966 | ||
GenBank mRNA | AK297026.1 | - | 6 | 706 | ||
BC003043.1 | - | 6 | 855 | 2822973 | ||
BC033104.1 | - | 6 | 913 | 4862014 | ||
M57567.1 | - | 6 | 873 |