Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145443.2 | NP_001138915.1 | 7 | 637 | ||
NM_001282630.2 | NP_001269559.1 | 7 | 750 | |||
NM_001314063.1 | NP_001300992.1 | 7 | 961 | |||
NM_001323016.1 | NP_001309945.1 | 7 | 637 | |||
NM_001323017.1 | NP_001309946.1 | 7 | 1011 | |||
NM_004566.3 | NP_004557.1 | 7 | 961 | |||
NR_136554.1 | 7 | 687 | ||||
XM_005252464.1 | XP_005252521.1 | 7 | 961 | |||
XM_011519493.1 | XP_011517795.1 | 7 | 961 | |||
XM_017016325.1 | XP_016871814.1 | 7 | 961 | |||
XM_017016326.1 | XP_016871815.1 | 6 | 904 | |||
XM_017016327.1 | XP_016871816.1 | 7 | 713 | |||
XM_017016328.1 | XP_016871817.1 | 7 | 961 | |||
XM_017016329.1 | XP_016871818.1 | 6 | 904 | |||
GenBank mRNA | AF056320.1 | - | 8 | 621 | ||
AF109735.1 | - | 7 | 691 | |||
AK131307.1 | - | 7 | 647 | |||
AK291263.1 | - | 7 | 906 | |||
AK303039.1 | - | 7 | 570 | |||
AK304450.1 | - | 7 | 682 | |||
BC037258.1 | - | 7 | 562 | |||
BC040482.1 | - | 7 | 906 | 5271907 | ||
D49817.1 | - | 7 | 595 | |||
L77662.1 | - | 7 | 915 |