Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK10 SIRNA Assays ›
Gene Symbol:
MAPK10
Gene Name:
mitogen-activated protein kinase 10
Gene Aliases:
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
Chromosome Location:
Chr.4: 86012296 - 86594117 on Build GRCh38
UniGene ID:
Hs.125503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
103462
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318067.1
NP_001304996.1
10
1127
NM_001318068.1
NP_001304997.1
10
1222
NM_001318069.1
NP_001304998.1
11
1692
NM_002753.4
NP_002744.1
11
1228
NM_138980.3
NP_620446.1
11
1719
NM_138982.3
NP_620448.1
11
1692
XM_005263129.2
XP_005263186.1
11
1789
XM_005263130.2
XP_005263187.1
11
1185
XM_005263131.2
XP_005263188.1
11
1586
XM_005263135.3
XP_005263192.1
11
1586
XM_006714268.2
XP_006714331.1
11
1459
XM_006714269.2
XP_006714332.1
11
1586
XM_011532117.2
XP_011530419.1
11
1292
XM_011532118.2
XP_011530420.1
11
1385
XM_011532120.2
XP_011530422.1
11
1355
XM_011532121.2
XP_011530423.1
11
1221
XM_017008419.1
XP_016863908.1
12
1482
XM_017008420.1
XP_016863909.1
11
1916
XM_017008421.1
XP_016863910.1
11
1512
XM_017008422.1
XP_016863911.1
8
940
XM_017008423.1
XP_016863912.1
11
1204
XM_017008424.1
XP_016863913.1
11
1539
XM_017008425.1
XP_016863914.1
11
1212
XM_017008426.1
XP_016863915.1
11
1540
XM_017008427.1
XP_016863916.1
11
1613
XM_017008428.1
XP_016863917.1
11
1355
XM_017008429.1
XP_016863918.1
11
1916
XM_017008430.1
XP_016863919.1
11
1294
XM_017008431.1
XP_016863920.1
11
1512
XM_017008432.1
XP_016863921.1
11
1185
XM_017008433.1
XP_016863922.1
10
1541
XM_017008434.1
XP_016863923.1
11
1569
XM_017008435.1
XP_016863924.1
11
1459
XM_017008436.1
XP_016863925.1
11
1943
XM_017008437.1
XP_016863926.1
11
1221
XM_017008438.1
XP_016863927.1
11
1212
XM_017008439.1
XP_016863928.1
11
1540
XM_017008440.1
XP_016863929.1
11
1613
XM_017008441.1
XP_016863930.1
10
1541
XM_017008442.1
XP_016863931.1
13
1934
XM_017008443.1
XP_016863932.1
13
1585
XM_017008444.1
XP_016863933.1
13
1907
XM_017008445.1
XP_016863934.1
10
1416
XM_017008446.1
XP_016863935.1
8
2108
XM_017008447.1
XP_016863936.1
10
1554
XM_017008448.1
XP_016863937.1
10
1527
XM_017008449.1
XP_016863938.1
10
1554
XM_017008450.1
XP_016863939.1
10
1527
XM_017008451.1
XP_016863940.1
10
1353
XM_017008452.1
XP_016863941.1
10
1554
GenBank mRNA
AK057723.1
-
6
1014
AK091104.1
-
3
958
AK124791.1
-
11
1944
AK225974.1
-
11
1273
AK294125.1
-
7
743
BC022492.1
-
11
1525
BC035057.1
-
11
1537
5261221
BC051731.1
-
11
1338
4838495
BC065516.1
-
6
689
5721954
U07620.1
-
11
1229
U34819.1
-
10
1073
U34820.1
-
10
1097
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012806, NM_001270556, XM_008769992, FQ211777, L27128, and others.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
protein phosphorylation
signal transduction
JNK cascade
JUN phosphorylation
response to light stimulus
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
JUN kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0