Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007118.3 | NP_009049.2 | 47 | 6710 | ||
NR_134469.1 | 47 | 6710 | ||||
XM_011514107.2 | XP_011512409.1 | 47 | 7285 | |||
XM_011514108.1 | XP_011512410.1 | 47 | 6584 | |||
XM_011514109.2 | XP_011512411.1 | 47 | 10362 | |||
XM_011514110.2 | XP_011512412.1 | 47 | 6820 | |||
XM_017009801.1 | XP_016865290.1 | 47 | 7062 | |||
XM_017009802.1 | XP_016865291.1 | 47 | 7062 | |||
XM_017009803.1 | XP_016865292.1 | 40 | 5331 | |||
GenBank mRNA | AB209754.1 | - | 42 | 5604 | ||
AF091395.1 | - | 47 | 6575 | |||
U42390.1 | - | 47 | 6575 |