Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK10 SIRNA Assays ›
Gene Symbol:
MAPK10
Gene Name:
mitogen-activated protein kinase 10
Gene Aliases:
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
Chromosome Location:
Chr.4: 86012296 - 86594117 on Build GRCh38
UniGene ID:
Hs.125503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
103540
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318067.1
NP_001304996.1
12
1363
NM_001318068.1
NP_001304997.1
12
1458
NM_001318069.1
NP_001304998.1
13
1928
NM_002753.4
NP_002744.1
13
1464
NM_138980.3
NP_620446.1
13
1955
NM_138982.3
NP_620448.1
13
1928
XM_005263129.2
XP_005263186.1
13
2025
XM_005263130.2
XP_005263187.1
13
1421
XM_005263131.2
XP_005263188.1
13
1822
XM_005263135.3
XP_005263192.1
13
1822
XM_006714268.2
XP_006714331.1
13
1695
XM_006714269.2
XP_006714332.1
13
1822
XM_011532117.2
XP_011530419.1
13
1528
XM_011532118.2
XP_011530420.1
13
1621
XM_011532120.2
XP_011530422.1
13
1591
XM_011532121.2
XP_011530423.1
13
1457
XM_017008419.1
XP_016863908.1
14
1718
XM_017008420.1
XP_016863909.1
13
2152
XM_017008421.1
XP_016863910.1
13
1748
XM_017008422.1
XP_016863911.1
10
1176
XM_017008423.1
XP_016863912.1
13
1440
XM_017008424.1
XP_016863913.1
13
1775
XM_017008425.1
XP_016863914.1
13
1448
XM_017008426.1
XP_016863915.1
13
1776
XM_017008427.1
XP_016863916.1
13
1849
XM_017008428.1
XP_016863917.1
13
1591
XM_017008429.1
XP_016863918.1
13
2152
XM_017008430.1
XP_016863919.1
13
1530
XM_017008431.1
XP_016863920.1
13
1748
XM_017008432.1
XP_016863921.1
13
1421
XM_017008433.1
XP_016863922.1
12
1777
XM_017008434.1
XP_016863923.1
13
1805
XM_017008435.1
XP_016863924.1
13
1695
XM_017008436.1
XP_016863925.1
13
2179
XM_017008437.1
XP_016863926.1
13
1457
XM_017008438.1
XP_016863927.1
13
1448
XM_017008439.1
XP_016863928.1
13
1776
XM_017008440.1
XP_016863929.1
13
1849
XM_017008441.1
XP_016863930.1
12
1777
XM_017008442.1
XP_016863931.1
15
2170
XM_017008443.1
XP_016863932.1
15
1821
XM_017008444.1
XP_016863933.1
15
2143
XM_017008445.1
XP_016863934.1
12
1652
XM_017008446.1
XP_016863935.1
10
2344
XM_017008447.1
XP_016863936.1
12
1790
XM_017008448.1
XP_016863937.1
12
1763
XM_017008449.1
XP_016863938.1
12
1790
XM_017008450.1
XP_016863939.1
12
1763
XM_017008451.1
XP_016863940.1
12
1589
XM_017008452.1
XP_016863941.1
12
1790
GenBank mRNA
AK057723.1
-
8
1250
AK091104.1
-
5
1194
AK124791.1
-
13
2180
AK225974.1
-
13
1509
AK294125.1
-
9
979
BC022492.1
-
13
1761
BC035057.1
-
13
1773
5261221
BC051731.1
-
13
1574
4838495
BC065516.1
-
8
925
5721954
U07620.1
-
13
1465
U34819.1
-
12
1309
U34820.1
-
12
1333
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
protein phosphorylation
signal transduction
JNK cascade
JUN phosphorylation
response to light stimulus
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
JUN kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-nglqg:80/10.239.166.193:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0