Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032294.2 | NP_115670.1 | 14 | 1452 | ||
NM_172206.1 | NP_757343.1 | 14 | 1398 | |||
NM_172207.2 | NP_757344.2 | 15 | 1566 | |||
XM_005256817.1 | XP_005256874.1 | 15 | 1566 | |||
XM_006721588.2 | XP_006721651.1 | 15 | 2126 | |||
XM_011524031.2 | XP_011522333.2 | 15 | 1685 | |||
XM_011524033.1 | XP_011522335.1 | 13 | 1372 | |||
XM_011524035.2 | XP_011522337.1 | 9 | 893 | |||
XM_017025219.1 | XP_016880708.1 | 14 | 1561 | |||
XM_017025220.1 | XP_016880709.1 | 14 | 2012 | |||
XM_017025221.1 | XP_016880710.1 | 7 | 634 | |||
GenBank mRNA | AF370377.1 | - | 1 | 1048 | ||
AF425232.1 | - | 14 | 1405 | |||
AF425301.1 | - | 14 | 1398 | |||
AL136576.1 | - | 14 | 1452 | |||
BC031647.1 | - | 15 | 1500 | 5175128 | ||
BC043487.1 | - | 14 | 1428 | 5751570 |