Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031741.3 | NP_001026911.1 | 5 | 662 | ||
NM_001304384.1 | NP_001291313.1 | 5 | 662 | |||
NM_152534.4 | NP_689747.3 | 27 | 2762 | |||
XM_006712997.3 | XP_006713060.1 | 27 | 2723 | |||
XM_006712998.2 | XP_006713061.1 | 27 | 2723 | |||
XM_006712999.3 | XP_006713062.1 | 27 | 2723 | |||
XM_006713001.3 | XP_006713064.1 | 27 | 2723 | |||
XM_017005761.1 | XP_016861250.1 | 26 | 2675 | |||
XM_017005762.1 | XP_016861251.1 | 28 | 2839 | |||
XM_017005763.1 | XP_016861252.1 | 27 | 2820 | |||
XM_017005764.1 | XP_016861253.1 | 26 | 2652 | |||
XM_017005765.1 | XP_016861254.1 | 26 | 2624 | |||
XM_017005766.1 | XP_016861255.1 | 26 | 2675 | |||
XM_017005767.1 | XP_016861256.1 | 25 | 2588 | |||
XM_017005768.1 | XP_016861257.1 | 26 | 2585 | |||
XM_017005769.1 | XP_016861258.1 | 27 | 2723 | |||
XM_017005770.1 | XP_016861259.1 | 26 | 2675 | |||
XM_017005771.1 | XP_016861260.1 | 26 | 2675 | |||
XM_017005772.1 | XP_016861261.1 | 25 | 2588 | |||
XM_017005773.1 | XP_016861262.1 | 27 | 2723 | |||
GenBank mRNA | AK057247.1 | - | 5 | 614 | ||
AK098832.1 | - | 8 | 845 | |||
BC045758.1 | - | 5 | 614 | 4821808 | ||
DQ104438.1 | - | 4 | 385 |