Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039366.1 | NP_001034455.1 | 4, 5 | 495 | ||
NM_001696.3 | NP_001687.1 | 5, 6 | 561 | |||
GenBank mRNA | AK294623.1 | - | 4, 5 | 422 | ||
AK312394.1 | - | 5, 6 | 482 | |||
BC004443.1 | - | 5, 6 | 463 | 2820930 | ||
BI546032.1 | - | 5, 6 | 513 | |||
BQ888820.1 | - | 4, 5 | 440 | |||
BT007128.1 | - | 5, 6 | 374 | |||
CK005516.1 | - | 5, 6 | 564 | |||
CR456385.1 | - | 5, 6 | 427 | |||
X71491.1 | - | 5, 6 | 430 | |||
X76228.1 | - | 5, 6 | 449 |