Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GALK2 SIRNA Assays ›
Gene Symbol:
GALK2
Gene Name:
galactokinase 2
Gene Aliases:
GK2
Chromosome Location:
Chr.15: 49155757 - 49367883 on Build GRCh38
UniGene ID:
Hs.122006
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
103624
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GALK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001556.2
NP_001001556.1
4, 5
570
NM_001289030.1
NP_001275959.1
5, 6
802
NM_001289031.1
NP_001275960.1
4, 5
655
NM_002044.3
NP_002035.1
4, 5
672
XM_005254279.3
XP_005254336.1
4, 5
461
XM_005254280.3
XP_005254337.1
4, 5
552
XM_005254284.3
XP_005254341.1
4, 5
552
XM_006720461.3
XP_006720524.1
4, 5
461
XM_006720462.3
XP_006720525.1
4, 5
442
XM_006720463.3
XP_006720526.1
4, 5
441
XM_006720464.3
XP_006720527.1
4, 5
441
XM_011521441.2
XP_011519743.1
5, 6
665
XM_017022062.1
XP_016877551.1
4, 5
442
XM_017022063.1
XP_016877552.1
4, 5
552
XM_017022064.1
XP_016877553.1
2, 3
518
XM_017022065.1
XP_016877554.1
5, 6
717
XM_017022066.1
XP_016877555.1
5, 6
738
GenBank mRNA
AF461816.1
-
4, 5
550
AK098049.1
-
5, 6
526
AK301361.1
-
4, 5
425
AK316440.1
-
5, 6
536
AL832745.1
-
4, 5
549
BC005141.2
-
4, 5
440
3504930
BQ921903.1
-
4, 5
383
BT006901.1
-
4, 5
365
EB386877.1
-
2, 3
201
M84443.1
-
4, 5
385
Back To Top
More Information
Panther Classification:
Molecular Function -
carbohydrate kinase
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
galactose metabolic process
carbohydrate phosphorylation
galactokinase activity
ATP binding
N-acetylgalactosamine kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0