Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317784.1 | NP_001304713.1 | 9 | 1487 | ||
NM_017988.5 | NP_060458.3 | 9 | 1639 | |||
XM_017019618.1 | XP_016875107.1 | 9 | 1487 | |||
XM_017019619.1 | XP_016875108.1 | 9 | 1639 | |||
XM_017019620.1 | XP_016875109.1 | 10 | 1626 | |||
GenBank mRNA | AB037781.1 | - | 6 | 793 | ||
AK000936.1 | - | 8 | 1245 | |||
AK024274.1 | - | 5 | 648 | |||
AK027551.1 | - | 5 | 705 | |||
AK125091.1 | - | 9 | 1624 | |||
AK292980.1 | - | 9 | 1413 | |||
BC063798.1 | - | 9 | 1252 | 6473586 |