Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006255.4 | NP_006246.2 | 10, 11 | 2087 | ||
XM_011536954.2 | XP_011535256.1 | 10, 11 | 1270 | |||
XM_011536955.1 | XP_011535257.1 | 10, 11 | 1203 | |||
XM_017021458.1 | XP_016876947.1 | 10, 11 | 1421 | |||
GenBank mRNA | AB451313.1 | - | 10, 11 | 1428 | ||
AK290183.1 | - | 10, 11 | 1806 | |||
AK296158.1 | - | 9, 10 | 1101 | |||
AK300864.1 | - | 2, 3 | 229 | |||
AX775931.1 | - | 10, 11 | 1163 | |||
AX775933.1 | - | 10, 11 | 1816 | |||
BC037268.1 | - | 10, 11 | 1816 | 4822159 | ||
M55284.1 | - | 10, 11 | 1591 |