Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323538.1 | NP_001310467.1 | 3, 4 | 335 | ||
NM_001761.2 | NP_001752.2 | 4, 5 | 439 | |||
XM_017023822.1 | XP_016879311.1 | 4, 5 | 599 | |||
XM_017023823.1 | XP_016879312.1 | 4, 5 | 599 | |||
GenBank mRNA | AB208829.1 | - | 3, 4 | 246 | ||
AK313371.1 | - | 4, 5 | 392 | |||
BC012349.1 | - | 4, 5 | 363 | 4634116 | ||
DA960397.1 | - | 3, 4 | 332 | |||
HY027359.1 | - | 4, 5 | 442 | |||
U17105.1 | - | 4, 5 | 364 | |||
Z36714.1 | - | 4, 5 | 394 |