Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000902.3 | NP_000893.2 | 17, 18 | 1773 | ||
NM_007287.2 | NP_009218.2 | 17, 18 | 1749 | |||
NM_007288.2 | NP_009219.2 | 17, 18 | 1795 | |||
NM_007289.2 | NP_009220.2 | 17, 18 | 1840 | |||
XM_006713646.3 | XP_006713709.1 | 17, 18 | 1787 | |||
XM_006713647.3 | XP_006713710.1 | 17, 18 | 4933 | |||
XM_011512855.2 | XP_011511157.1 | 17, 18 | 1864 | |||
XM_011512856.2 | XP_011511158.1 | 17, 18 | 1791 | |||
XM_011512857.2 | XP_011511159.1 | 17, 18 | 1762 | |||
GenBank mRNA | AK291761.1 | - | 17, 18 | 1864 | ||
BC101632.1 | - | 17, 18 | 1731 | 8069138 | ||
BC101658.1 | - | 17, 18 | 1729 | 8069164 | ||
BC143465.1 | - | 17, 18 | 1731 | 9051973 | ||
BX647134.1 | - | 17, 18 | 1744 | |||
J03779.1 | - | 16, 17 | 1664 | |||
X07166.1 | - | 16, 17 | 1649 | |||
Y00811.1 | - | 17, 18 | 1788 |