Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001871.2 | NP_001862.2 | 6, 7 | 607 | ||
GenBank mRNA | AJ224866.1 | - | 6, 7 | 589 | ||
AK313940.1 | - | 6, 7 | 595 | |||
BC015338.1 | - | 6, 7 | 588 | 4422620 | ||
BI770145.1 | - | 6, 7 | 607 | |||
BT009910.1 | - | 6, 7 | 584 | |||
M81057.1 | - | 6, 7 | 607 |