Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001977.3 | NP_001968.3 | 14, 15 | 2501 | ||
XM_017007877.1 | XP_016863366.1 | 11, 12 | 1179 | |||
GenBank mRNA | AK313412.1 | - | 15, 16 | 2163 | ||
AM888273.1 | - | 14, 15 | 2501 | |||
AM888274.1 | - | 14, 15 | 2501 | |||
AM888275.1 | - | 14, 15 | 2501 | |||
AM888276.1 | - | 14, 15 | 2501 | |||
AM888277.1 | - | 14, 15 | 2501 | |||
AM888278.1 | - | 14, 15 | 2501 | |||
AM888279.1 | - | 14, 15 | 2501 | |||
AM888280.1 | - | 14, 15 | 2501 | |||
AM888281.1 | - | 14, 15 | 2501 | |||
AM888282.1 | - | 14, 15 | 2501 | |||
AM888283.1 | - | 14, 15 | 2501 | |||
BC094770.1 | - | 15, 16 | 2482 | 30528477 | ||
BX641012.1 | - | 11, 12 | 1668 | |||
L12468.1 | - | 15, 16 | 2405 | |||
L14721.1 | - | 15, 16 | 2242 |